Learn More
Midwest Scientific Accucold Undercounter All Refrigerator | Undercounter 5.5 Cu. Ft. Refrigerator | Laboratory Undercounter Refrigerator
SDP
Catalog No. NC9086220
Encompass
Automatic defrost saves on time and maintenance, while Accucold's unique design simplifies clean-up. A "hidden" evaporator set behind the rear wall creates a seamless interior, permitting to wipe down the back without having to get between crevices or touch any machinery. 5.5 cubic feet of all refrigerator All refrigerator (no freezer section) Full automatic defrost with adjustable thermostat. Adjustable wire shelves. Height-33.5" X Width-23.63" X Depth-23.5"
Catalog No. NC9086220 Supplier Midwest Scientific Supplier No. AT5AR
Automatic defrost saves on time and maintenance, while Accucold's unique design simplifies clean-up. A "hidden" evaporator set behind the rear wall creates a seamless interior, permitting to wipe down the back without having to get between crevices or touch any machinery. 5.5 cubic feet of all refrigerator All refrigerator (no freezer section) Full automatic defrost with adjustable thermostat. Adjustable wire shelves. Height-33.5" X Width-23.63" X Depth-23.5"