Learn More
PHCbi Undercounter refrigerator
Catalog No. 502720031
Encompass_Preferred
PHCbi brand 5.0 cu.ft. (142 L) purpose built undercounter freezer which requires less energy than traditional undercounter medical refrigerators. This model feaure a glass door for conventient viweing of invetnory without unneccesary door opening and is designed to maintain the high perforance necessary for valuable inventory using inverter compressor speed technology for optimum control. ENERGY STAR (R) CertfiedTemperature Range+2°C to +8°C
Catalog No. 50-272-0031 Supplier PHCbi Supplier No. PRL5181GWPA
PHCbi brand 5.0 cu.ft. (142 L) purpose built undercounter freezer which requires less energy than traditional undercounter medical refrigerators. This model feaure a glass door for conventient viweing of invetnory without unneccesary door opening and is designed to maintain the high perforance necessary for valuable inventory using inverter compressor speed technology for optimum control. ENERGY STAR (R) CertfiedTemperature Range+2°C to +8°C