Learn More
PHCbi Undercounter Freezer
Catalog No. 502720032
Encompass_Preferred
PHCbi brand 5.0 cu.ft. (142 L) purpose built undercounter freezer which requires less energy than traditional undercounter medical refrigerators. This model is designed to maintain the high perforance necessary for valuable inventory using inverter compressor speed technology for optimum control. ENERGY STAR (R) CertfiedTemperature Range-15°C to -25°C
Catalog No. 50-272-0032 Supplier PHCbi Supplier No. PFL5181WPA
Add to Cart
Add to Cart
PHCbi brand 5.0 cu.ft. (142 L) purpose built undercounter freezer which requires less energy than traditional undercounter medical refrigerators. This model is designed to maintain the high perforance necessary for valuable inventory using inverter compressor speed technology for optimum control. ENERGY STAR (R) CertfiedTemperature Range-15°C to -25°C