Learn More
Grainger Underground Enclosure Cover 18-1/8 L
Catalog No. 502764256
Encompass_Preferred
The Quazite Underground Enclosure Cover is a heavy duty, non-metallic enclosure cover designed for underground electrical, cable TV and telecom applications. It is made from polymer concrete for durability and has a load rating of 22,500 lbs. The dimensions are 24"L x 24"W x 8"H with a 6"H dome. It features pry slots for easy removal and has a gray finish. The cover weighs approximately 65 lbs. This enclosure cover provides secure underground access while protecting electrical and telecom connections from the elements.
Catalog No. 50-276-4256 Supplier Grainger Supplier No. 4AWX9
The Quazite Underground Enclosure Cover is a heavy duty, non-metallic enclosure cover designed for underground electrical, cable TV and telecom applications. It is made from polymer concrete for durability and has a load rating of 22,500 lbs. The dimensions are 24"L x 24"W x 8"H with a 6"H dome. It features pry slots for easy removal and has a gray finish. The cover weighs approximately 65 lbs. This enclosure cover provides secure underground access while protecting electrical and telecom connections from the elements.