Learn More
Poltex Undermount Bulk Bin
SDP
Catalog No. 21377577
Change view
Click to view available options
Mounting Type:
Magnetic Mount
Screw Mount
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting Type
21-377-577 Magnetic Mount
21-377-578 Screw Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-377-577 Supplier Poltex Supplier No. UNDRBULK1M
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
Bulk bin that mounts under laboratory bench shelves and protrudes into usable position.
Mounts under laboratory bench shelves but protrudes into usable position
Durable PETG construction
Choose from:
  • Powerful magnet mount for mounting to metal shelves
  • Screw mount to mount to wood/melamine shelves
Specifications
Product Type Organizer
Autoclavable Not Autoclavable
Color Clear
Mounting Type Magnetic Mount
No. of Doors None
No. of Shelves None
Material PETG