Learn More
Poltex Undermount Cord Keeper
SDP
Catalog No. 21377581
Change view
Click to view available options
Mounting Type:
Magnetic Mount
Catalog No. Mounting Type
21-377-581 Magnetic Mount
1 options
Catalog No. 21-377-581 Supplier Poltex Supplier No. UNDRCORDM
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
Mounts under laboratory bench shelves to utilize space then protrudes into usable position.
Mounts under laboratory bench shelves
Protrudes into usable position
Durable PETG construction
Choose from:
  • Magnet mount for mounting to metal shelves
  • Screw mount to mount to wood/melamine shelves
Specifications
Product Type Organizer
Autoclavable Not Autoclavable
Mounting Type Magnetic Mount
Color Clear
No. of Doors None
No. of Shelves None
Material PETG