Learn More
Poltex Undermount Double Bulk Bin
SDP
Catalog No. 21377579
Change view
Click to view available options
Mounting Type:
Magnetic Mount
Screw Mount
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting Type
21-377-579 Magnetic Mount
21-377-580 Screw Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-377-579 Supplier Poltex Supplier No. UNDRBULK2M
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
Double bulk bin that mounts under laboratory bench shelves and protrudes into usable position.
Mounts under laboratory bench shelves but protrudes into usable position
Durable PETG construction
Choose from:
  • Powerful magnet mount for mounting to metal shelves
  • Screw mount to mount to wood/melamine shelves
Specifications
Product Type Organizer
Color Clear
Autoclavable Not Autoclavable
Mounting Type Magnetic Mount
No. of Shelves None
No. of Doors None
Material PETG