Learn More
Poltex Undermount Serological Pipette Holder with 2 Pockets
SDP
Catalog No. 21377597
Change view
Click to view available options
Mounting Type:
Magnetic Mount
Screw Mount
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting Type
21-377-597 Magnetic Mount
21-377-598 Screw Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-377-597 Supplier Poltex Supplier No. UNDRSRPIP2M
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
Mounts under laboratory bench shelves to utilize space then protrudes into usable position.
Mounts under laboratory bench shelves
Protrudes into usable position
Durable PETG construction
Choose from:
  • Magnet mount for mounting to metal shelves
  • Screw mount to mount to wood/melamine shelves
Specifications
Product Type Organizer
Autoclavable Not Autoclavable
Mounting Type Magnetic Mount
No. of Shelves None
Color Clear
No. of Doors None
Material PETG