Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex LLC Undermount Universal Pipette Hanger, 2 Positions
SDP

Catalog No. 01259313
Change view
Click to view available options
Mounting:
Magnet Mount
Screw Mount
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
01-259-313 Magnet Mount
01-259-307 Screw Mount
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-259-313 Supplier Poltex LLC Supplier No. UNDRPIPJ2M
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Part of the Poltex UnderMount line of organizers. Made in the USA.

  • The UnderMount line of organizers mounts under laboratory bench shelves to utilize the space that every lab has, but is normally not used
  • Item mounts under the shelf, but protrudes into usable position for the end-user
  • Fits any size pipettor due to simple cradle design
  • Choose from powerful magnet mount for mounting to metal shelves, or screw mount to mount to wood/melamine shelves
  • Durable and cleanable PETG construction

Specifications

Autoclavable Not Autoclavable
Color Clear
Holds Pipettes
Material PETG
Positions 1
Shape J-shaped
Stackable No
Sterility Non-sterile
For Use With (Equipment) Universal Pipette
Mounting Magnet Mount
Dimensions (L x W x H) 5.8 x 6.6 x 5.4 in.
Length (English) 5.8 in.
Height (English) 5.4 in.
Product Type Pipette Holder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.