Learn More
Med Vet International Super Absorbent Underpads, 23"X36", 10/Pack
Catalog No. 502829897
Encompass_Preferred
The WeekEnder Premium Disposable Puppy Pads.*Advanced Supercore thermo-bonded core.*Wicks moisture away from the surface and locks fluid in within minutes.*Super absorbent--1 pad absorbs what used to require 3 or more.*Super strong to reduce tearing and resulting damage to floors.*Superior odor control.*Folded over edge for better containment.*Soft non-woven top sheet across the pad.*Ultra absorbent polymer is spread throughout the entire core for maximum absorbency.*Top sheet is fused to the core so it does not bunch up while in use.*Waterproof polyethylene backing protects underlying surfaces.
Catalog No. 50-282-9897 Supplier Med Vet International Supplier No. SUPERPAD
The WeekEnder Premium Disposable Puppy Pads.*Advanced Supercore thermo-bonded core.*Wicks moisture away from the surface and locks fluid in within minutes.*Super absorbent--1 pad absorbs what used to require 3 or more.*Super strong to reduce tearing and resulting damage to floors.*Superior odor control.*Folded over edge for better containment.*Soft non-woven top sheet across the pad.*Ultra absorbent polymer is spread throughout the entire core for maximum absorbency.*Top sheet is fused to the core so it does not bunch up while in use.*Waterproof polyethylene backing protects underlying surfaces.