Learn More
Med Vet International Disposable Underpad TENA Extra 23 X 36 Inch Polymer Light Absorbency, 355, 25/Bag
Catalog No. 503044090
Encompass_Preferred
Tena underpads for bed or chair protection are designed to cover mattresses in order to absorb leakages, reduce odors and maintain dryness They consist of a polyethylene layer combined with sealed edges to ensure leakage protection The highly absorbent material is evenly distributed throughout the core
Catalog No. 50-304-4090 Supplier Med Vet International Supplier No. 465698BG
Tena underpads for bed or chair protection are designed to cover mattresses in order to absorb leakages, reduce odors and maintain dryness They consist of a polyethylene layer combined with sealed edges to ensure leakage protection The highly absorbent material is evenly distributed throughout the core