Learn More
Med Vet International McKesson Ultra Breathable Heavy Absorbency Low Air Loss Disposable Underpad, 23 x 36 Inch, UPHV2336, 10/Bag
Catalog No. 503044267
Encompass_Preferred
Disposable Underpad McKesson Ultra Breathable 23 X 36 Inch Fluff / Polymer Heavy Absorbency Designed for use on airflow therapy beds, featuring a fluff core with superior mat configuration Super-absorbent polymer locks wetness away for advanced odor control pH of urine is neutralized for the sake of skin wellness Soft, nonwoven bonded topsheet promotes rapid fluid acquisition Backsheet is breathable for a cool, comfortable design Heat-sealed, nonwoven edges feel gentle on skin
Catalog No. 50-304-4267 Supplier Med Vet International Supplier No. 724054BG
Disposable Underpad McKesson Ultra Breathable 23 X 36 Inch Fluff / Polymer Heavy Absorbency Designed for use on airflow therapy beds, featuring a fluff core with superior mat configuration Super-absorbent polymer locks wetness away for advanced odor control pH of urine is neutralized for the sake of skin wellness Soft, nonwoven bonded topsheet promotes rapid fluid acquisition Backsheet is breathable for a cool, comfortable design Heat-sealed, nonwoven edges feel gentle on skin