Learn More
Med Vet International McKesson Classic Light Absorbency Disposable Underpad, 23 x 36 Inch, UPLT2336, 10/Bag
Catalog No. 503045011
Encompass_Preferred
Disposable Underpad McKesson Classic Plus 23 X 36 Inch Fluff Mat Light Absorbency Offers protection for general procedures and wound care Fluff/polymer core provides super mat configuration Nonwoven bonded topsheet promotes rapid fluid acquisition Heat-sealed, nonwoven edges feel gentle on skin
Catalog No. 50-304-5011 Supplier Med Vet International Supplier No. 724033BG
Disposable Underpad McKesson Classic Plus 23 X 36 Inch Fluff Mat Light Absorbency Offers protection for general procedures and wound care Fluff/polymer core provides super mat configuration Nonwoven bonded topsheet promotes rapid fluid acquisition Heat-sealed, nonwoven edges feel gentle on skin