Learn More
Med Vet International McKesson Ultimate Breathable Disposable Underpads, Maximum Protection, 24" x 36", UPMX2436, 5/Bag
Catalog No. 503042951
Encompass_Preferred
Disposable Underpad McKesson Ultimate Breathable 24 X 36 Inch Fluff / Polymer Heavy Absorbency McKesson underpads with super absorbent polymer lock in wetness to keep you dry and protect surfaces from damage They have a breathable backing to reduce skin irritations The non-woven, heat-sealed edges enhance user comfort while reducing tears A fluff core and mat configuration on the breathable underpads reduces odors The latex-free material is suitable for all skin types 24 inches x 36 inches size provides protection to a larger area
Catalog No. 50-304-2951 Supplier Med Vet International Supplier No. 1075428BG
Disposable Underpad McKesson Ultimate Breathable 24 X 36 Inch Fluff / Polymer Heavy Absorbency McKesson underpads with super absorbent polymer lock in wetness to keep you dry and protect surfaces from damage They have a breathable backing to reduce skin irritations The non-woven, heat-sealed edges enhance user comfort while reducing tears A fluff core and mat configuration on the breathable underpads reduces odors The latex-free material is suitable for all skin types 24 inches x 36 inches size provides protection to a larger area