Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Understanding Blood Type Interactions Through Simulated Blood Typing Kit
GREENER_CHOICESCI_ED

Catalog No. S96790R Shop All Aldon Products
Change view
Click to view available options
Product Type:
Simulated Blood Typing Kit
Simulated Blood Typing Refill
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S96790R Simulated Blood Typing Refill
S96790 Simulated Blood Typing Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S96790R Supplier Innovating Science™ Supplier No. IS3103REF
Only null left
Add to Cart
Add to Cart

Includes

  • Four simulated blood samples
  • One bottle each anti-A, anti-B and anti-Rh serum
  • 40 blood typing trays

Use for deeper understanding of the importance of antigen/antibody interactions.

Use simulated blood to determine the ABO/Rh blood type of four individuals, one in need of transfusion. The result determines which of three potential donors would provide the best match for the patient in need.

  • Provides simulation of blood typing procedure
  • No toothpicks, stirring or waiting for results
  • Hands-on activities
  • Kit contains materials for 10 groups

Kit uses simulated blood only.

Specifications

Color White
Grade 7 to 14
Age 13+ years
For Use With (Application) For simulated blood kit
Includes Teacher's manual, student guide, 40 blood typing trays, 4 simulated blood samples, donor no. 1, donor no. 2, donor no. 3, patient, 1 bottle simulated anti-a serum, 1 bottle simulated anti-b serum, 1 bottle simulated anti-rh serum
Quantity 1
Class Size 10 Lab Groups
Product Type Simulated Blood Typing Refill
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.