Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Understanding pH Lab Investigation
SCI_ED

Catalog No. s67400
Change view
Click to view available options
:
Understand pH Lab Investigation
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S67400 Understand pH Lab Investigation
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S67400 Supplier Neo/Sci Corporation Supplier No. 201743

Discover the crucial role pH plays in the balance of life.

Gain an understanding of the importance of pH in biological chemical reactions and in the well-being of cells, organs and overall health.

  • Learn the range of indicators required to determine acidity and alkalinity
  • Measure the pH values of several colorless solutions
  • Classify unknown solutions based on their relative acid and base strengths
  • Observe the outcomes of mixing specific acids and bases
  • See the color change of acid/base indicators
  • Explain the relationships between acids, bases and salts
  • Test how antacids neutralize stomach acid

Order Info

Class size: up to 40 students.

Specifications

For Use With (Application) Importance of ph in biological chemical reactions
Weight (English) 3 lb.
Weight (Metric) 1.3 kg
Class Size 40 Students
Product Type Laboratory Exercise
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.