Learn More
Grainger Undr Flr Elc Heat Systm 10 ft.x36 240V
Catalog No. 502749106
Encompass_Preferred
The Grainger 14C462 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA 48Y frame with a 1.15 service factor. The motor is totally enclosed, fan cooled (TEFC) with a cast iron body. It has a C-face mounting style and measures approximately 9-1/2" long by 7-3/4" wide by 8-3/4" high. The motor weighs approximately 35 pounds and has a 1/2" diameter shaft with a 1/4" keyway and threaded conduit box. It can be wired for high or low voltage and is rated for continuous duty applications.
Catalog No. 50-274-9106 Supplier Grainger Supplier No. 14C462
The Grainger 14C462 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA 48Y frame with a 1.15 service factor. The motor is totally enclosed, fan cooled (TEFC) with a cast iron body. It has a C-face mounting style and measures approximately 9-1/2" long by 7-3/4" wide by 8-3/4" high. The motor weighs approximately 35 pounds and has a 1/2" diameter shaft with a 1/4" keyway and threaded conduit box. It can be wired for high or low voltage and is rated for continuous duty applications.