Learn More
Grainger ThermoSoft Under Floor Electric Heating System, 120V, 30 Sq. Ft.
Catalog No. 502752885
Encompass_Preferred
This professional-grade, 4-in-1 radiant floor heating underlayment pad provides heating, insulation, noise reduction, and moisture abatement in one 1/8 in. thick layer. Designed for indoor use under laminate, wood, vinyl, and carpet flooring, it utilizes patented FiberThermics® technology to ensure even heat distribution through uniform wire spacing. The system eliminates the need for cement installation, making it suitable for floating floors.
  • Features a 98% recycled fiber core with chemical and abrasion-resistant insulation
  • Provides 10 W/sq. ft. output for efficient radiant heating
  • Includes a thermal cut-off safety feature
  • Listed to UL 1693 and CSA 22.2 No. 130-2003 standards
  • Offers 70 dB Impact Isolation Class (IIC) and 65 dB Sound Transmission Class (STC) ratings
  • 10-year manufacturer warranty
Catalog No. 50-275-2885 Supplier Grainger Supplier No. 14C458
This professional-grade, 4-in-1 radiant floor heating underlayment pad provides heating, insulation, noise reduction, and moisture abatement in one 1/8 in. thick layer. Designed for indoor use under laminate, wood, vinyl, and carpet flooring, it utilizes patented FiberThermics® technology to ensure even heat distribution through uniform wire spacing. The system eliminates the need for cement installation, making it suitable for floating floors.
  • Features a 98% recycled fiber core with chemical and abrasion-resistant insulation
  • Provides 10 W/sq. ft. output for efficient radiant heating
  • Includes a thermal cut-off safety feature
  • Listed to UL 1693 and CSA 22.2 No. 130-2003 standards
  • Offers 70 dB Impact Isolation Class (IIC) and 65 dB Sound Transmission Class (STC) ratings
  • 10-year manufacturer warranty