Learn More
Grainger Undr Flr Elc Heat Systm 20 ft.x36 240V
Catalog No. 502750517
Encompass_Preferred
The Grainger Approved Under Floor Electric Heat System is 20 feet long by 36 inches wide and operates on 240 volts. It provides radiant floor heating for areas up to 72 square feet. The system features a thin, flexible heating wire that is sandwiched between layers of fiberglass mesh. It can be installed under many types of flooring including tile, stone, laminate, hardwood and carpet. The wire heats to a uniform 105°F across the entire mat. This underfloor heating mat comes with a built-in thermostat to control the temperature.
Catalog No. 50-275-0517 Supplier Grainger Supplier No. 14C463
The Grainger Approved Under Floor Electric Heat System is 20 feet long by 36 inches wide and operates on 240 volts. It provides radiant floor heating for areas up to 72 square feet. The system features a thin, flexible heating wire that is sandwiched between layers of fiberglass mesh. It can be installed under many types of flooring including tile, stone, laminate, hardwood and carpet. The wire heats to a uniform 105°F across the entire mat. This underfloor heating mat comes with a built-in thermostat to control the temperature.