Learn More
Grainger Undr Flr Elc Heat Systm 40 ft.x18 240V
Catalog No. 502749058
Encompass_Preferred
The Grainger 14C450 is a 14-inch diameter, 450 CFM, direct drive exhaust fan. It features a thermally protected PSC motor and is designed for general ventilation applications. The fan has a galvanized steel housing with baked enamel finish and includes adjustable motor mounts. It requires 115V power and is ETL listed for safety. The 14C450 can provide powerful airflow for industrial and commercial ventilation needs. With proper installation, this exhaust fan will provide years of reliable operation.
Catalog No. 50-274-9058 Supplier Grainger Supplier No. 14C450
The Grainger 14C450 is a 14-inch diameter, 450 CFM, direct drive exhaust fan. It features a thermally protected PSC motor and is designed for general ventilation applications. The fan has a galvanized steel housing with baked enamel finish and includes adjustable motor mounts. It requires 115V power and is ETL listed for safety. The 14C450 can provide powerful airflow for industrial and commercial ventilation needs. With proper installation, this exhaust fan will provide years of reliable operation.