Learn More
Med Vet International ReliaMed Underpad, Reusable, Polyester, 36in x 54in, ZRUP3654R, 10/Case
Catalog No. 503042961
Encompass_Preferred
Reusable Underpad Cardinal Health™ Essentials 36 X 54 Inch Polyester / Rayon Moderate Absorbency ReliaMed reusable underpads have a PVC-coated backsheet that protects surfaces from stains The backsheet material is skid resistant to prevent the underpads from slipping The underpads are made of high-quality fabric that retains its efficiency after many washes The top sheet wicks moisture away from the skin to keep you dry and protected The waterproof underpads have a poly/rayon core that offers a moderate absorbency and helps eliminate odors
Catalog No. 50-304-2961 Supplier Med Vet International Supplier No. 1120201CS
Reusable Underpad Cardinal Health™ Essentials 36 X 54 Inch Polyester / Rayon Moderate Absorbency ReliaMed reusable underpads have a PVC-coated backsheet that protects surfaces from stains The backsheet material is skid resistant to prevent the underpads from slipping The underpads are made of high-quality fabric that retains its efficiency after many washes The top sheet wicks moisture away from the skin to keep you dry and protected The waterproof underpads have a poly/rayon core that offers a moderate absorbency and helps eliminate odors