Learn More
Med Vet International Disposable Underpad TENA Regular 17 X 24 Inch Fluff Light Absorbency, 350, 12/Case
Catalog No. 503043389
Encompass_Preferred
TENA underpads soak up leakages to keep surfaces dry and minimize odors A fluffy core filled with absorbent polymers guards against light incontinence A sealed, waterproof lining locks wetness away A soft, non-woven outer layer offers comfort and a gentle feel on the skin A latex-free construction increases safety for patients and caregivers TENA blue underpads are completely disposable for convenience and hassle-free cleanups
Catalog No. 50-304-3389 Supplier Med Vet International Supplier No. 450009CS
TENA underpads soak up leakages to keep surfaces dry and minimize odors A fluffy core filled with absorbent polymers guards against light incontinence A sealed, waterproof lining locks wetness away A soft, non-woven outer layer offers comfort and a gentle feel on the skin A latex-free construction increases safety for patients and caregivers TENA blue underpads are completely disposable for convenience and hassle-free cleanups