Learn More
Med Vet International Disposable Underpad McKesson Ultra 36 X 36 Inch Fluff / Polymer Heavy Absorbency, UPHV3636, 5/Bag, 10 Bags
Catalog No. 503045014
Encompass_Preferred
Super-absorbent polymer locks wetness away for advanced odor control pH of urine is neutralized to assist skin wellness Fluff core provides super mat configuration Nonwoven bonded topsheet promotes rapid fluid acquisition Heat-sealed, nonwoven edges feel gentle on skin
Catalog No. 50-304-5014 Supplier Med Vet International Supplier No. 724038CS
Super-absorbent polymer locks wetness away for advanced odor control pH of urine is neutralized to assist skin wellness Fluff core provides super mat configuration Nonwoven bonded topsheet promotes rapid fluid acquisition Heat-sealed, nonwoven edges feel gentle on skin