Learn More
Med Vet International Reusable Underpad Beck's Classic 36 X 54 Inch Polyester / Rayon Moderate Absorbency, 7155GRN-PB, Each
Catalog No. 503043271
Encompass_Preferred
Underpads are constructed with 3 layers of soft and thick quilting combined with a Vintex vinyl barrier and a special five-thread safety stitch for extra durability 80% cotton / 20% polyester Quilted to a blended poly/rayon soaker Knitted vinyl barrier Green
Catalog No. 50-304-3271 Supplier Med Vet International Supplier No. 1125494EA
Underpads are constructed with 3 layers of soft and thick quilting combined with a Vintex vinyl barrier and a special five-thread safety stitch for extra durability 80% cotton / 20% polyester Quilted to a blended poly/rayon soaker Knitted vinyl barrier Green