Learn More
Med Vet International Tranquility Heavy Duty Disposable Underpad, 30 x 36in, 2088, 10/Bag
Catalog No. 503044235
Encompass_Preferred
Disposable Underpad Tranquility Heavy Duty 30 X 36 Inch Super Absorbent Polymer Heavy Absorbency Provides 7½ square feet (30 x 36 inch) of protection and can absorb up to 1 quart of fluid Extra-strong, cloth-like backsheet is slip resistant and resists tearing
Catalog No. 50-304-4235 Supplier Med Vet International Supplier No. 798791BG
Disposable Underpad Tranquility Heavy Duty 30 X 36 Inch Super Absorbent Polymer Heavy Absorbency Provides 7½ square feet (30 x 36 inch) of protection and can absorb up to 1 quart of fluid Extra-strong, cloth-like backsheet is slip resistant and resists tearing