Learn More
Med Vet International Disposable Underpad McKesson Super 23 X 36 Inch Fluff / Polymer Moderate Absorbency, UPMD2336, 10/Bag, 15 Bags
Catalog No. 503044627
Encompass_Preferred
Super-absorbent polymer locks wetness away for advanced odor control pH of urine is neutralized to assist skin wellness Fluff core provides super mat configuration Nonwoven bonded topsheet promotes rapid fluid acquisition Heat-sealed, nonwoven edges feel gentle on skin
Catalog No. 50-304-4627 Supplier Med Vet International Supplier No. 724035CS
Super-absorbent polymer locks wetness away for advanced odor control pH of urine is neutralized to assist skin wellness Fluff core provides super mat configuration Nonwoven bonded topsheet promotes rapid fluid acquisition Heat-sealed, nonwoven edges feel gentle on skin