Learn More
Med Vet International Disposable Underpad Simplicity™ Basic 17 X 24 Inch Fluff Light Absorbency, 7105A, 300/Case
Catalog No. 503044400
Encompass_Preferred
17 x 24 inch pad Light absorbency Fluff absorbent core Polypropylene backsheet protects against leakage and resists melting Spunbound non-woven top sheet allows fluid to pass quickly into the absorbent core
Catalog No. 50-304-4400 Supplier Med Vet International Supplier No. 1210421CS
17 x 24 inch pad Light absorbency Fluff absorbent core Polypropylene backsheet protects against leakage and resists melting Spunbound non-woven top sheet allows fluid to pass quickly into the absorbent core