Learn More
Med Vet International Disposable Underpad, WINGS™, 23 x 24 Inch, Fluff, Light Absorbency, 7134, 200/Case
Catalog No. 503046368
Encompass_Preferred
Spunbound non-woven top sheet allows fluid to pass quickly into the absorbent core Polypropylene backsheet protects against leakage and resists melting Fluff absorbent core
Catalog No. 50-304-6368 Supplier Med Vet International Supplier No. 1256032CS
Spunbound non-woven top sheet allows fluid to pass quickly into the absorbent core Polypropylene backsheet protects against leakage and resists melting Fluff absorbent core