Learn More
Selleck Chemical LLC Unesbulin PTC596-2mg
SDP
Catalog No. 502389859
Encompass_Preferred
Unesbulin (PTC596) is a second-generation BMI-1 inhibitor that accelerates BMI-1 degradation. PTC596 downregulates MCL-1 and induces p53-independent mitochondrial apoptosis. IC50 values at 72 hours ranged from 68 to 340 nM in mantle cell lymphoma (MCL) cell lines.
Catalog No. 50-238-9859 Supplier Selleck Chemical LLC Supplier No. S88202MG
Unesbulin (PTC596) is a second-generation BMI-1 inhibitor that accelerates BMI-1 degradation. PTC596 downregulates MCL-1 and induces p53-independent mitochondrial apoptosis. IC50 values at 72 hours ranged from 68 to 340 nM in mantle cell lymphoma (MCL) cell lines.