Learn More
Aviva Systems Biology Corp UNG Antibody - C-terminal region (ARP61205_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61205_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against UNG. It was validated on Western Blot. . Alternate names: DGU, DKFZp781L1143, HIGM4, UDG, UNG1, UNG15, UNG2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-287-294