Learn More
Enterprise Technology Solutions Unger Cleaner Kit,f/Outdoor Glass,Deionizing,Large,39' Pole,Gray
SDP
Catalog No. 5000829666
Encompass_Preferred
Hydropower Ultra Large Carbon Kit offers a fast, safe and efficient way to professionally clean glass surfaces outdoors. One-stage water tank uses deionizing technology to produce 100 percent pure water for more effective cleaning, rinsing and more. Modern water system design produces 30 percent more pure water output and leaves a spot-free finish. 39' nLITE carbon 24K pole easily cleans up to 44' from ground level to eliminate the need for ladders. FloWater technology ensures even, more efficient water distribution throughout the system, improving results and increasing its lifespan. Safe, self-locking FastLock mechanism uses a pressure release valves for quick-and-easy opening. Kit includes HydroPower window cleaning equipment, nLITE six-section carbon 24K pole, nLITE master/extension connection, DuroFlex hose, nLITE carrying bag, water flow regular, power brush and more.
Catalog No. 50-008-29666 Supplier Enterprise Technology Solutions Supplier No. UNGUNLK5
Hydropower Ultra Large Carbon Kit offers a fast, safe and efficient way to professionally clean glass surfaces outdoors. One-stage water tank uses deionizing technology to produce 100 percent pure water for more effective cleaning, rinsing and more. Modern water system design produces 30 percent more pure water output and leaves a spot-free finish. 39' nLITE carbon 24K pole easily cleans up to 44' from ground level to eliminate the need for ladders. FloWater technology ensures even, more efficient water distribution throughout the system, improving results and increasing its lifespan. Safe, self-locking FastLock mechanism uses a pressure release valves for quick-and-easy opening. Kit includes HydroPower window cleaning equipment, nLITE six-section carbon 24K pole, nLITE master/extension connection, DuroFlex hose, nLITE carrying bag, water flow regular, power brush and more.