Learn More
Enterprise Technology Solutions Unger Cleaning Kit,f/Windows,Profes.,76 Piece,Nylon/Plastic,BK/GN
SDP
Catalog No. 5000789789
Encompass_Preferred
ErgoTec Professional Window Cleaning Kit with 76 pieces makes window-washing jobs easier than ever. Included cleaning tools help you achieve both a professional-grade clean and ergonomic comfort during use. Microfiber StripWasher Sleeve, attached by hook-and-loop closure, can survive up to 500 wash cycles and absorbs up to six times its own weight in liquid. Stainless steel S-Channels secure rubber squeegee inserts for effective, streak-free squeegee performance and professional finish. ErgoTec scraper's reversible carbon-steel blade scrapes paint and tape with its razor edge and cement and stucco with its blunt edge. OptiLoc 2-Section Pole made from aluminum easily fits various tools using ErgoTec Locking Cones to limit ladder use.
Catalog No. 50-007-89789 Supplier Enterprise Technology Solutions Supplier No. UNGETSET
ErgoTec Professional Window Cleaning Kit with 76 pieces makes window-washing jobs easier than ever. Included cleaning tools help you achieve both a professional-grade clean and ergonomic comfort during use. Microfiber StripWasher Sleeve, attached by hook-and-loop closure, can survive up to 500 wash cycles and absorbs up to six times its own weight in liquid. Stainless steel S-Channels secure rubber squeegee inserts for effective, streak-free squeegee performance and professional finish. ErgoTec scraper's reversible carbon-steel blade scrapes paint and tape with its razor edge and cement and stucco with its blunt edge. OptiLoc 2-Section Pole made from aluminum easily fits various tools using ErgoTec Locking Cones to limit ladder use.