Learn More
Enterprise Technology Solutions Unger MicroWipe™ 2000 Microfiber Cloth, Medium-Duty, 16"x16", Green
SDP
Catalog No. 5000828855
Encompass_Preferred
MicroWipe 2000 Medium-Duty Microfiber Cloth traps dirt and absorbs liquids for effective cleaning. Ideal for medium-duty tasks, it provides superior absorbency and cleaning power on a variety of surfaces. Whether you're cleaning glass, stainless steel or other surfaces, this microfiber cloth ensures a streak-free, polished finish every time. Its large, 16" x 16" size covers more surface area, making cleaning faster and more efficient. Microfiber cleaning cloths reduce surface debris while lowering bacteria levels by more than 96%, according to laboratory tests. Mold, bacteria and soil are removed and captured thanks to its unique construction. With color-coded options, using this system also supports cross-contamination prevention strategies, making it ideal for use in environments where hygiene is critical. Cloth is machine-washable and reusable for long-lasting performance.
Catalog No. 50-008-28855 Supplier Enterprise Technology Solutions Supplier No. UNGMB400
MicroWipe 2000 Medium-Duty Microfiber Cloth traps dirt and absorbs liquids for effective cleaning. Ideal for medium-duty tasks, it provides superior absorbency and cleaning power on a variety of surfaces. Whether you're cleaning glass, stainless steel or other surfaces, this microfiber cloth ensures a streak-free, polished finish every time. Its large, 16" x 16" size covers more surface area, making cleaning faster and more efficient. Microfiber cleaning cloths reduce surface debris while lowering bacteria levels by more than 96%, according to laboratory tests. Mold, bacteria and soil are removed and captured thanks to its unique construction. With color-coded options, using this system also supports cross-contamination prevention strategies, making it ideal for use in environments where hygiene is critical. Cloth is machine-washable and reusable for long-lasting performance.