Learn More
Enterprise Technology Solutions Unger Squeegee Blade, 12", 12/PK, Black
SDP
Supplier:  Enterprise Technology Solutions UNGRT30
Encompass_Preferred
12" squeegee blade is coated with talcum to keep it fresh and ready to use for high-quality results. Soft rubber construction conforms to slightly uneven surfaces to glide smoothly with streak-free performance. Natural molded rubber provides a sharp, long-lasting edge to thoroughly move viscous substances. Squeegee blade is perfect for use in colder working conditions.
Catalog No. 50-008-29752