Learn More
Enterprise Technology Solutions Unger Trigger Grip, 32", Magnetic Tip, 5/CT, Green/Silver
SDP
Catalog No. 5000829004
Encompass_Preferred
Nifty Nabber offers an ergonomically designed grip, magnetic tip, and smooth trigger-pull action to increase comfort and efficiency. Grab and hold for hard-to-reach areas or items. Access hard-to-reach items with a 360-degree rotating head. Lock head securely in place by pulling trigger and rotating head to desired position. Head locks every 90 degrees. Grab and hold even the smallest items with the claw's magnetic tip. Wide-claw design opens to 5" to make it easy to grab items with large diameters. The shaft is designed to decrease fatigue and make waist-high and overhead use easy. Grip is made with glass-reinforced plastic for increased durability. Shaft is made of durable, lightweight aluminum.
Catalog No. 50-008-29004 Supplier Enterprise Technology Solutions Supplier No. UNGNT080CT
Nifty Nabber offers an ergonomically designed grip, magnetic tip, and smooth trigger-pull action to increase comfort and efficiency. Grab and hold for hard-to-reach areas or items. Access hard-to-reach items with a 360-degree rotating head. Lock head securely in place by pulling trigger and rotating head to desired position. Head locks every 90 degrees. Grab and hold even the smallest items with the claw's magnetic tip. Wide-claw design opens to 5" to make it easy to grab items with large diameters. The shaft is designed to decrease fatigue and make waist-high and overhead use easy. Grip is made with glass-reinforced plastic for increased durability. Shaft is made of durable, lightweight aluminum.