Learn More
Organomation Unheated 12 position N-EVAP nitrogen evaporator
SDP
Supplier:  Organomation 11155O
Encompass_Preferred
The unheated 12 position N-EVAP (cat# 11155-O) has proven to be an extremely durable, time-tested, and highly affordable member of Organomation’s N-EVAP line. These economical units provide controlled concentration of sample vials through the application of nitrogen gas within ambient temperatures. The N-EVAP’s circular rotating design permits each sample to be accessible from the front of the instrument for easy insertion and retrieval. The chrome plated needle valves are individually milled from solid brass, enabling precise, uniform adjustments for your nitrogen blow down. This promotes efficient and safe sample reduction with minimal nitrogen waste. At each individual sample position, the gas flow can be controlled via a precision needle valve. This is ideal for the simultaneous evaporation of samples with varying evaporation end points. The standard sample tray allows any test tube between 10-30 mm OD to be processed.
Catalog No. NC2301755