Learn More
Organomation Unheated 24 position MICROVAP Small Vial Evaporator
SDP
Supplier:  Organomation 11824O
Encompass_Preferred
The unheated 24 position MICROVAP small vial evaporator is intended for concentrating small volume, small batch samples, such as concentrations of groups of samples in microcentrifuge tubes and GC vials. It offers an effective and compact solution to evaporation of samples for life sciences and pharmaceutical laboratories. Each MICROVAP uses minimal hood space while providing ambient temperature for heat sensitive solvents. The 24 position MICROVAP comes standard with stainless steel luer lock hubs and 4-inch x 19-gauge stainless steel needles for easy removal. Up to 24 samples can be accommodated at once, and the instrument comes customized for one sample size with an outside diameter of 10-17 mm. If multiple sample sizes will be used with the instrument, additional sample blocks can be purchased for each additional sample size.
Catalog No. NC2301743