Learn More
Organomation Unheated 45 Position N-EVAP
SDP
Catalog No. NC2301759
Encompass_Preferred
Our unheated 45-position nitrogen evaporator (Catalog # 11645-O) offers an ambient temperature solution for heat sensitive solvents. It has the largest number of sample positions in our N-EVAP line. The sample holder has dual circular rings. The outer ring has 30 positions and can accommodate samples with an outside diameter of 10-19 mm The inner ring has 15 positions and can accommodate samples with an outside diameter of 16-30 mm and a minimum height of 70 mm. This time-tested, durable system provides the least expensive price per position of all our N-EVAPs. The evaporative system, along with the standard flow control and pressure reducing regulator, enables the operator to achieve the desired level of concentration with efficient nitrogen usage. Our unique dual band spring hoist assembly ensures easy and effective operator use.
Catalog No. NC2301759 Supplier Organomation Supplier No. 11645O
Add to Cart
Add to Cart
Request a Quote
Our unheated 45-position nitrogen evaporator (Catalog # 11645-O) offers an ambient temperature solution for heat sensitive solvents. It has the largest number of sample positions in our N-EVAP line. The sample holder has dual circular rings. The outer ring has 30 positions and can accommodate samples with an outside diameter of 10-19 mm The inner ring has 15 positions and can accommodate samples with an outside diameter of 16-30 mm and a minimum height of 70 mm. This time-tested, durable system provides the least expensive price per position of all our N-EVAPs. The evaporative system, along with the standard flow control and pressure reducing regulator, enables the operator to achieve the desired level of concentration with efficient nitrogen usage. Our unique dual band spring hoist assembly ensures easy and effective operator use.