Learn More
OHAUS™ Uni Block
SCI_ED
Catalog No. S27298 Shop All OHAUS Products
Change view
Click to view available options
For Use With (Equipment):
28 mm Vials
Catalog No. For Use With (Equipment)
S27298 28 mm Vials
1 options
Catalog No. S27298 Supplier OHAUS™ Supplier No. 30500585/FSE
Add to Cart
Add to Cart
Hot plates and stirrers accessory
Holds test tubes or vials
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 28 mm Vials
Type Uni Block
Material Metal