Learn More
OHAUS™ Uni Block
Catalog No. 01922030 Shop All OHAUS Products
Change view
Click to view available options
For Use With (Equipment):
12 mm Test Tubes
12 mm Vials
15 mm Vials
16 mm Test Tubes
17 mm Vials
20 mm Test Tubes
21 mm Vials
25 mm Test Tubes
28 mm Vials
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
01-922-030 21 mm Vials
01-922-008 12 mm Test Tubes
01-922-009 16 mm Test Tubes
01-922-010 20 mm Test Tubes
01-922-011 25 mm Test Tubes
01-922-027 12 mm Vials
01-922-028 15 mm Vials
01-922-029 17 mm Vials
01-922-031 28 mm Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 01-922-030 Supplier OHAUS™ Supplier No. 30500584
Only null left
Add to Cart
Add to Cart
Hot plates and stirrers accessory
Holds test tubes or vials
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 21 mm Vials
Type Uni Block
Material Metal