Learn More
Research Products International Corp Universal Flask Tray for Orbital and Shaking Water Bath
SDP
Catalog No. 502229531
Encompass
For use with the Orbital and Shaking Water Bath.
Tray holds 26 x 25ml; 26 x 50ml; 15 x 100ml; 11 x 250ml; 6 x 500ml; or 3 x 1L.
Catalog No. 50-222-9531 Supplier Research Products International Corp Supplier No. 246607
For use with the Orbital and Shaking Water Bath.
Tray holds 26 x 25ml; 26 x 50ml; 15 x 100ml; 11 x 250ml; 6 x 500ml; or 3 x 1L.