Learn More
Med Vet International Uni-Pole Clamping IV Pole, J1576A, Each
Catalog No. 5000585512
Encompass_Preferred
The Uni-Pole IV pole clamping system clamps on about anything like a surgery table rail or other flat surfaces. The IV pole is made of chromed steel construction and screws into the adjustable black aluminum mounting clamp. • IV pole adjusts from 18" to 36" high • Adjustable clamp mounts on flat surfaces or round pole up to 1.25" thick • 2 ram horns IV bag holder
Catalog No. 50-005-85512 Supplier Med Vet International Supplier No. JOR1576A
The Uni-Pole IV pole clamping system clamps on about anything like a surgery table rail or other flat surfaces. The IV pole is made of chromed steel construction and screws into the adjustable black aluminum mounting clamp. • IV pole adjusts from 18" to 36" high • Adjustable clamp mounts on flat surfaces or round pole up to 1.25" thick • 2 ram horns IV bag holder