Learn More
Molecular Dimensions UNI-PUCK STARTER KIT
SDP
Catalog No. 501038605
Encompass
Uni-Puck Starter Kit Contents of Starter Kit: 7 x Uni-pucks, 1 x Puck-Shelved Shipping Cane, 1 x Angled Cryo-Tongs, 1 x Puck Separator Tools, 1 x Puck Wand, 1 x Puck Dewar Loading Tool Set and 1 x Puck Loading Dewar with Lid.
Catalog No. 50-103-8605 Supplier Molecular Dimensions Supplier No. MD7600
Add to Cart
Add to Cart
Request a Quote
Uni-Puck Starter Kit Contents of Starter Kit: 7 x Uni-pucks, 1 x Puck-Shelved Shipping Cane, 1 x Angled Cryo-Tongs, 1 x Puck Separator Tools, 1 x Puck Wand, 1 x Puck Dewar Loading Tool Set and 1 x Puck Loading Dewar with Lid.