Learn More
Analytical Sales & Services 11mm, 12 x 32 Clear Plastic Crimp/Snap MicroVials with 100µL - 300µL Glass Inserts , 100/Pk
Catalog No. 50450131
Encompass
Uni-Vial 11mm Crimp Snap Plast
Catalog No. 50-450-131 Supplier Analytical Sales & Services Supplier No. 11241
Uni-Vial 11mm Crimp Snap Plast