Learn More
Electron Microscopy Sciences Uniband Gripper
SDP
Catalog No. 5029561
Encompass_Preferred
Unique Uni-Band Gripper is made of 300 Series Memory Stainless Steel which is harder than titanium. Perfect gripper for handling specimen mount stubs.
Catalog No. 50-295-61 Supplier Electron Microscopy Sciences Supplier No. 76806
Unique Uni-Band Gripper is made of 300 Series Memory Stainless Steel which is harder than titanium. Perfect gripper for handling specimen mount stubs.