Learn More
Electron Microscopy Sciences MicroPoint™ Uniband LA-BS Bandage Cutter, 125 mm, 125 mm
SDP
Catalog No. 5027248
Encompass_Preferred
MicroPoint scissors can be used for suture or tissue and they are manufactured without rings which eliminates the problems with right handed or left handed users. Scissors cut by shearing and have three times the shearing angle to reduce "tissue crushing. They last far longer than ordinary scissors.
O.R. Castro-Viejo quality at a floor-grade price.
They offer consistent cutting at the tip.
The blades cannot work loose and the contact between the blades remains constant.
Adjustable shearing contact.
All MicroPoint™ Scissors are guaranteed for life against rust, breakage and rivet loosening.
OAL: Curved
Blade: 7 mm
Blade Size: 125 mm
Catalog No. 50-272-48 Supplier Electron Microscopy Sciences Supplier No. 77915BS
MicroPoint scissors can be used for suture or tissue and they are manufactured without rings which eliminates the problems with right handed or left handed users. Scissors cut by shearing and have three times the shearing angle to reduce "tissue crushing. They last far longer than ordinary scissors.
O.R. Castro-Viejo quality at a floor-grade price.
They offer consistent cutting at the tip.
The blades cannot work loose and the contact between the blades remains constant.
Adjustable shearing contact.
All MicroPoint™ Scissors are guaranteed for life against rust, breakage and rivet loosening.
OAL: Curved
Blade: 7 mm
Blade Size: 125 mm