Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Uniblue A Sodium Salt (Tech.), 80%, Thermo Scientific™

Catalog No. AC422420010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC422420010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC422420010 Supplier Thermo Scientific Chemicals Supplier No. 422420010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14541-90-3
Molecular Formula C22H16N2NaO7S2
Molecular Weight (g/mol) 507.487
InChI Key YBKLUTQGHNVYTN-UHFFFAOYSA-N
PubChem CID 71308378
IUPAC Name 1-amino-4-(3-ethenylsulfonylanilino)-9,10-dioxoanthracene-2-sulfonic acid;sodium
SMILES C=CS(=O)(=O)C1=CC=CC(=C1)NC2=CC(=C(C3=C2C(=O)C4=CC=CC=C4C3=O)N)S(=O)(=O)O.[Na]

Specifications

Infrared Spectrum Authentic
Formula Weight 506.48
Percent Purity ≥75%
Grade Technical
Suitability (for use as a protein stain in electrophoresis) passes
Color Blue
Quantity 1 g
Physical Form Fine Powder
Chemical Name or Material Uniblue A sodium salt, tech., 80%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.