Learn More
Beckman Coulter UniCel DxL 600 Access Immunoassay System
Catalog No. 21403556 Shop All Beckman Coulter Products
Change view
Click to view available options
Includes:
System Only
System with Spot B
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
21-403-556 System Only
21-403-557 System with Spot B
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-403-556 Supplier Beckman Coulter Supplier No. A45867
Only null left
Add to Cart
Add to Cart
Request a Quote
The UniCel DxI 600 Access Immunoassay System helps laboratories running 40,000 to 80,000 tests per year meet peak workload demands.
Built for scalability
  • Systems feature standardized test menus, assay protocols, instrument processes and reference ranges. Together, these features minimize training requirements and streamline inventory management tasks for labor cost savings, while providing consistent results across the healthcare delivery network
  • Results correlate across platforms for consistency in patient care
  • Continuous loading of STAT and routine samples address peak workflow requirements
Highly flexible
  • System releases the primary tube after creating an aliquot, making the system available for other uses
  • Refrigerated compartment holds 50 reagent packs in any configuration, allowing you to customize the system menu to match your testing needs
  • Performs up to 200 tests per hour
  • Broad assay test menu and simple operation give you the ability to expand in-house services
  • Capable of integrating with automation and IT systems
Optimized for the needs of technologists
  • Load reagents and consumables without pausing the instrument
  • Load samples and walk away to perform higher value work and return to results
  • Maximize walk-away time with large onboard supply capacity
  • View monitor and indicator lights to see which supplies need replenishment
  • Remotely diagnose analyzer with optional PROService
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Beckman Coulter Analyzers
Throughput 200 Tests/hr.
Includes System Only