Description
Built for scalability
- Systems feature standardized test menus, assay protocols, instrument processes and reference ranges. Together, these features minimize training requirements and streamline inventory management tasks for labor cost savings, while providing consistent results across the healthcare delivery network
- Results correlate across platforms for consistency in patient care
- Continuous loading of STAT and routine samples address peak workflow requirements
Highly flexible
- System releases the primary tube after creating an aliquot, making the system available for other uses
- Refrigerated compartment holds 50 reagent packs in any configuration, allowing you to customize the system menu to match your testing needs
- Performs up to 200 tests per hour
- Broad assay test menu and simple operation give you the ability to expand in-house services
- Capable of integrating with automation and IT systems
Optimized for the needs of technologists
- Load reagents and consumables without pausing the instrument
- Load samples and walk away to perform higher value work and return to results
- Maximize walk-away time with large onboard supply capacity
- View monitor and indicator lights to see which supplies need replenishment
- Remotely diagnose analyzer with optional PROService
Specifications
Specifications
| For Use With (Equipment) | Beckman Coulter Analyzers |
| Throughput | 200 Tests/hr. |
| Includes | System with Spot B |