Learn More
Apexbio Technology LLC Uniconazole 83657-22-1 10g
SDP
Catalog No. 5000088282
Encompass_Preferred
Uniconazole (CAS 83657-22-1) is a small-molecule inhibitor of cytochrome P450 707A enzymes (CYP707As) which are involved in the catabolism of abscisic acid (ABA) via 8 -hydroxylation In in vitro assays uniconazole demonstrated potent inhibition of CYP707A3 activity with a reported Ki of 68 nM surpassing other plant growth retardants such as paclobutrazol and tetcyclacis In Arabidopsis models uniconazole treatment elevated endogenous ABA levels and enhanced drought tolerance effects that were independent of gibberellin-mediated pathways Uniconazole is therefore utilized in plant biology research to study ABA metabolism and stress responses
Catalog No. 50-000-88282 Supplier Apexbio Technology LLC Supplier No. C507610000
Uniconazole (CAS 83657-22-1) is a small-molecule inhibitor of cytochrome P450 707A enzymes (CYP707As) which are involved in the catabolism of abscisic acid (ABA) via 8 -hydroxylation In in vitro assays uniconazole demonstrated potent inhibition of CYP707A3 activity with a reported Ki of 68 nM surpassing other plant growth retardants such as paclobutrazol and tetcyclacis In Arabidopsis models uniconazole treatment elevated endogenous ABA levels and enhanced drought tolerance effects that were independent of gibberellin-mediated pathways Uniconazole is therefore utilized in plant biology research to study ABA metabolism and stress responses